Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615587 MIM: 609288 | ||||||||||||||||||||
Literature Links: |
NUP188 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUP188 - nucleoporin 188 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3GLB2 - SH3 domain containing GRB2 like endophilin B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287045.1 | 1054 | Missense Mutation | AGC,TGC | S,C 271 | NP_001273974.1 | |
NM_001287046.1 | 1054 | Missense Mutation | AGC,TGC | S,C 267 | NP_001273975.1 | |
NM_020145.3 | 1054 | Missense Mutation | AGC,TGC | S,C 267 | NP_064530.1 | |
XM_005252098.2 | 1054 | Missense Mutation | AGC,TGC | S,C 267 | XP_005252155.1 | |
XM_005252100.2 | 1054 | Missense Mutation | AGC,TGC | S,C 246 | XP_005252157.1 | |
XM_005252101.3 | 1054 | UTR 5 | XP_005252158.1 | |||
XM_006717188.2 | 1054 | Missense Mutation | AGC,TGC | S,C 295 | XP_006717251.1 | |
XM_006717189.2 | 1054 | Missense Mutation | AGC,TGC | S,C 291 | XP_006717252.1 | |
XM_006717190.1 | 1054 | Missense Mutation | AGC,TGC | S,C 295 | XP_006717253.1 | |
XM_006717191.1 | 1054 | Missense Mutation | AGC,TGC | S,C 271 | XP_006717254.1 |