Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103950 MIM: 604874 | ||||||||||||||||||||||||||||||||
Literature Links: |
A2M PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
A2M - alpha-2-macroglobulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000014.4 | 4549 | Intron | NP_000005.2 | |||
XM_006719056.2 | 4549 | Silent Mutation | TGC,TGT | C,C 1486 | XP_006719119.1 |
A2M-AS1 - A2M antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLRG1 - killer cell lectin like receptor G1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005810.3 | 4549 | Intron | NP_005801.3 | |||
XM_017018682.1 | 4549 | Intron | XP_016874171.1 | |||
XM_017018683.1 | 4549 | Intron | XP_016874172.1 | |||
XM_017018684.1 | 4549 | Intron | XP_016874173.1 | |||
XM_017018685.1 | 4549 | Intron | XP_016874174.1 |
LINC00612 - long intergenic non-protein coding RNA 612 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |