Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5D2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYB5D2 - cytochrome b5 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254755.1 | 251 | Intron | NP_001241684.1 | |||
NM_001254756.1 | 251 | Intron | NP_001241685.1 | |||
NM_144611.3 | 251 | Intron | NP_653212.1 | |||
XM_006721447.2 | 251 | UTR 5 | XP_006721510.2 | |||
XM_011523649.2 | 251 | Intron | XP_011521951.1 | |||
XM_017024180.1 | 251 | Intron | XP_016879669.1 | |||
XM_017024181.1 | 251 | Intron | XP_016879670.1 | |||
XM_017024182.1 | 251 | UTR 5 | XP_016879671.1 | |||
XM_017024183.1 | 251 | Intron | XP_016879672.1 |
ZZEF1 - zinc finger ZZ-type and EF-hand domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |