Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMKK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMKK1 - calcium/calmodulin dependent protein kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032294.2 | 2608 | UTR 3 | NP_115670.1 | |||
NM_172206.1 | 2608 | UTR 3 | NP_757343.1 | |||
NM_172207.2 | 2608 | Intron | NP_757344.2 | |||
XM_005256817.1 | 2608 | UTR 3 | XP_005256874.1 | |||
XM_006721588.2 | 2608 | UTR 3 | XP_006721651.1 | |||
XM_011524031.2 | 2608 | Intron | XP_011522333.2 | |||
XM_011524032.2 | 2608 | Intron | XP_011522334.1 | |||
XM_011524033.1 | 2608 | UTR 3 | XP_011522335.1 | |||
XM_011524035.2 | 2608 | UTR 3 | XP_011522337.1 | |||
XM_017025219.1 | 2608 | UTR 3 | XP_016880708.1 | |||
XM_017025220.1 | 2608 | UTR 3 | XP_016880709.1 | |||
XM_017025221.1 | 2608 | UTR 3 | XP_016880710.1 | |||
XM_017025222.1 | 2608 | Intron | XP_016880711.1 |