Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCGTTCTCACGACCTCTGCCCT[A/G]GCACACGAGCTTCGAGTTATAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612959 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ESRP1 - epithelial splicing regulatory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034915.2 | Intron | NP_001030087.2 | ||||
NM_001122825.1 | Intron | NP_001116297.1 | ||||
NM_001122826.1 | Intron | NP_001116298.1 | ||||
NM_001122827.1 | Intron | NP_001116299.1 | ||||
NM_017697.3 | Intron | NP_060167.2 | ||||
XM_005250991.3 | Intron | XP_005251048.1 | ||||
XM_005250992.3 | Intron | XP_005251049.1 |
LOC100288748 - uncharacterized LOC100288748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |