Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CREB5 - cAMP responsive element binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011666.2 | Intron | NP_001011666.1 | ||||
NM_004904.3 | Intron | NP_004895.2 | ||||
NM_182898.3 | Intron | NP_878901.2 | ||||
NM_182899.4 | Intron | NP_878902.2 | ||||
XM_005249906.1 | Intron | XP_005249963.1 | ||||
XM_011515618.2 | Intron | XP_011513920.1 | ||||
XM_017012806.1 | Intron | XP_016868295.1 | ||||
XM_017012807.1 | Intron | XP_016868296.1 | ||||
XM_017012808.1 | Intron | XP_016868297.1 | ||||
XM_017012809.1 | Intron | XP_016868298.1 | ||||
XM_017012810.1 | Intron | XP_016868299.1 |
Set Membership: |
HapMap |