Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610849 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTLL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TTLL6 - tubulin tyrosine ligase like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130918.1 | Intron | NP_001124390.1 | ||||
NM_173623.3 | Intron | NP_775894.2 | ||||
XM_017024491.1 | Intron | XP_016879980.1 | ||||
XM_017024492.1 | Intron | XP_016879981.1 | ||||
XM_017024493.1 | Intron | XP_016879982.1 | ||||
XM_017024494.1 | Intron | XP_016879983.1 | ||||
XM_017024495.1 | Intron | XP_016879984.1 | ||||
XM_017024496.1 | Intron | XP_016879985.1 | ||||
XM_017024497.1 | Intron | XP_016879986.1 |
Set Membership: |
HapMap |