Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609231 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUDT7 - nudix hydrolase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105663.2 | Intron | NP_001099133.1 | ||||
NM_001243657.1 | Intron | NP_001230586.1 | ||||
NM_001243660.1 | Intron | NP_001230589.1 | ||||
NM_001243661.1 | Intron | NP_001230590.1 | ||||
XM_011523032.2 | Intron | XP_011521334.1 | ||||
XM_011523033.2 | Intron | XP_011521335.1 | ||||
XM_017023170.1 | Intron | XP_016878659.1 |
Set Membership: |
HapMap |