Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606623 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSD17B6 - hydroxysteroid 17-beta dehydrogenase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003725.3 | Intron | NP_003716.2 | ||||
XM_005269207.1 | Intron | XP_005269264.1 | ||||
XM_005269208.1 | Intron | XP_005269265.1 | ||||
XM_005269209.1 | Intron | XP_005269266.1 | ||||
XM_006719672.1 | Intron | XP_006719735.1 | ||||
XM_011538925.1 | Intron | XP_011537227.1 | ||||
XM_011538926.1 | Intron | XP_011537228.1 | ||||
XM_011538927.1 | Intron | XP_011537229.1 |
Set Membership: |
HapMap |