Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609736 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC88A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC88A - coiled-coil domain containing 88A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135597.1 | 7914 | UTR 3 | NP_001129069.1 | |||
NM_001254943.1 | 7914 | UTR 3 | NP_001241872.1 | |||
NM_018084.4 | 7914 | UTR 3 | NP_060554.3 | |||
XM_005264418.4 | 7914 | Intron | XP_005264475.1 | |||
XM_005264421.2 | 7914 | UTR 3 | XP_005264478.1 | |||
XM_005264426.2 | 7914 | Intron | XP_005264483.1 | |||
XM_011532965.2 | 7914 | UTR 3 | XP_011531267.1 | |||
XM_011532966.2 | 7914 | Intron | XP_011531268.1 | |||
XM_011532967.2 | 7914 | Intron | XP_011531269.1 | |||
XM_011532968.2 | 7914 | UTR 3 | XP_011531270.1 | |||
XM_017004476.1 | 7914 | Intron | XP_016859965.1 | |||
XM_017004477.1 | 7914 | Intron | XP_016859966.1 | |||
XM_017004478.1 | 7914 | Intron | XP_016859967.1 | |||
XM_017004479.1 | 7914 | Intron | XP_016859968.1 | |||
XM_017004480.1 | 7914 | Intron | XP_016859969.1 | |||
XM_017004481.1 | 7914 | Intron | XP_016859970.1 |
PRORSD1P - prolyl-tRNA synthetase associated domain containing 1, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |