Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143170 MIM: 601498 MIM: 601646 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MEA1 - male-enhanced antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318942.1 | Intron | NP_001305871.1 | ||||
NM_001318943.1 | Intron | NP_001305872.1 | ||||
NM_014623.3 | Intron | NP_055438.1 | ||||
XM_005249121.3 | Intron | XP_005249178.1 | ||||
XM_017010868.1 | Intron | XP_016866357.1 |
PEX6 - peroxisomal biogenesis factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP2R5D - protein phosphatase 2 regulatory subunit B'delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270476.1 | Intron | NP_001257405.1 | ||||
NM_006245.3 | Intron | NP_006236.1 | ||||
NM_180976.2 | Intron | NP_851307.1 | ||||
NM_180977.2 | Intron | NP_851308.1 |
Set Membership: |
HapMap |