Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603028 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF175 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF175 - ring finger protein 175 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173662.2 | Intron | NP_775933.1 | ||||
XM_005262938.3 | Intron | XP_005262995.1 | ||||
XM_005262939.3 | Intron | XP_005262996.1 | ||||
XM_005262940.4 | Intron | XP_005262997.1 | ||||
XM_011531879.2 | Intron | XP_011530181.1 | ||||
XM_011531881.2 | Intron | XP_011530183.1 | ||||
XM_011531882.2 | Intron | XP_011530184.1 | ||||
XM_011531883.2 | Intron | XP_011530185.1 | ||||
XM_017008047.1 | Intron | XP_016863536.1 |
TLR2 - toll like receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |