Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603344 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
G3BP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
G3BP2 - G3BP stress granule assembly factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012297.4 | Intron | NP_036429.2 | ||||
NM_203504.2 | Intron | NP_987100.1 | ||||
NM_203505.2 | Intron | NP_987101.1 | ||||
XM_005263382.2 | Intron | XP_005263439.1 | ||||
XM_005263383.3 | Intron | XP_005263440.1 | ||||
XM_011532441.2 | Intron | XP_011530743.1 | ||||
XM_017008876.1 | Intron | XP_016864365.1 | ||||
XM_017008877.1 | Intron | XP_016864366.1 | ||||
XM_017008878.1 | Intron | XP_016864367.1 | ||||
XM_017008879.1 | Intron | XP_016864368.1 |
USO1 - USO1 vesicle transport factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290049.1 | Intron | NP_001276978.1 | ||||
NM_003715.3 | Intron | NP_003706.2 | ||||
XM_006714396.3 | Intron | XP_006714459.1 | ||||
XM_006714397.3 | Intron | XP_006714460.1 |
Set Membership: |
HapMap |