Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606975 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COG3 - component of oligomeric golgi complex 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ERICH6B - glutamate rich 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182542.2 | 1805 | Missense Mutation | CGC,TGC | R,C 565 | NP_872348.2 | |
XM_011534965.2 | 1805 | Missense Mutation | CGC,TGC | R,C 589 | XP_011533267.1 | |
XM_017020418.1 | 1805 | Missense Mutation | CGC,TGC | R,C 565 | XP_016875907.1 |