Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAGTTTATGGACCTAGTGTACT[A/G]GCAGTGAAAAAAATTTCATATCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612175 MIM: 607865 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAB39L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAB39L - calcium binding protein 39 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETDB2 - SET domain bifurcated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160308.2 | Intron | NP_001153780.1 | ||||
NM_001320699.1 | Intron | NP_001307628.1 | ||||
NM_031915.2 | Intron | NP_114121.2 |
SETDB2-PHF11 - SETDB2-PHF11 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320727.1 | Intron | NP_001307656.1 |
Set Membership: |
HapMap |