Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECT2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ECT2L - epithelial cell transforming 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077706.2 | Intron | NP_001071174.1 | ||||
NM_001195037.2 | Intron | NP_001181966.1 | ||||
XM_006715472.3 | Intron | XP_006715535.1 | ||||
XM_011535795.2 | Intron | XP_011534097.1 | ||||
XM_011535797.2 | Intron | XP_011534099.1 | ||||
XM_017010828.1 | Intron | XP_016866317.1 | ||||
XM_017010829.1 | Intron | XP_016866318.1 | ||||
XM_017010830.1 | Intron | XP_016866319.1 |