Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEBP2 - heme binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NHSL1 - NHS like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144060.1 | 9014 | UTR 3 | NP_001137532.1 | |||
NM_020464.1 | 9014 | UTR 3 | NP_065197.1 | |||
XM_005267062.4 | 9014 | UTR 3 | XP_005267119.1 | |||
XM_011535965.2 | 9014 | UTR 3 | XP_011534267.1 | |||
XM_011535966.2 | 9014 | UTR 3 | XP_011534268.1 | |||
XM_011535967.2 | 9014 | UTR 3 | XP_011534269.1 | |||
XM_011535968.2 | 9014 | UTR 3 | XP_011534270.1 | |||
XM_011535969.2 | 9014 | UTR 3 | XP_011534271.1 | |||
XM_011535970.2 | 9014 | UTR 3 | XP_011534272.1 | |||
XM_011535971.2 | 9014 | UTR 3 | XP_011534273.1 | |||
XM_011535972.2 | 9014 | UTR 3 | XP_011534274.1 | |||
XM_011535973.2 | 9014 | UTR 3 | XP_011534275.1 | |||
XM_011535974.2 | 9014 | UTR 3 | XP_011534276.1 | |||
XM_011535976.1 | 9014 | UTR 3 | XP_011534278.1 | |||
XM_017011086.1 | 9014 | UTR 3 | XP_016866575.1 | |||
XM_017011087.1 | 9014 | UTR 3 | XP_016866576.1 | |||
XM_017011088.1 | 9014 | UTR 3 | XP_016866577.1 | |||
XM_017011089.1 | 9014 | UTR 3 | XP_016866578.1 |