Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATCCCTTTAAAATTCATACATTA[C/T]TTAGGACCCCAAAGGGCTTTTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604645 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928326 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928326 - uncharacterized LOC101928326 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE7B - phosphodiesterase 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018945.3 | Intron | NP_061818.1 | ||||
XM_005266931.3 | Intron | XP_005266988.1 | ||||
XM_011535755.2 | Intron | XP_011534057.1 | ||||
XM_017010741.1 | Intron | XP_016866230.1 |
Set Membership: |
HapMap |