Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615757 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIZ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIZ - kizuna centrosomal protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163022.1 | Intron | NP_001156494.1 | ||||
NM_001163023.1 | Intron | NP_001156495.1 | ||||
NM_001276389.1 | Intron | NP_001263318.1 | ||||
NM_018474.4 | Intron | NP_060944.3 | ||||
XM_011529296.2 | Intron | XP_011527598.1 | ||||
XM_011529297.2 | Intron | XP_011527599.1 | ||||
XM_011529298.2 | Intron | XP_011527600.1 | ||||
XM_011529299.2 | Intron | XP_011527601.1 | ||||
XM_017027951.1 | Intron | XP_016883440.1 | ||||
XM_017027952.1 | Intron | XP_016883441.1 | ||||
XM_017027953.1 | Intron | XP_016883442.1 | ||||
XM_017027954.1 | Intron | XP_016883443.1 |