Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCATCTGCAGACATTTCTAGGTC[C/T]CCAATGCTGAGCAAATATATCTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610222 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987276 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC107987276 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017028183.1 | Intron | XP_016883672.1 |
RIN2 - Ras and Rab interactor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242581.1 | Intron | NP_001229510.1 | ||||
NM_018993.3 | Intron | NP_061866.1 | ||||
XM_005260731.2 | Intron | XP_005260788.1 | ||||
XM_005260733.2 | Intron | XP_005260790.1 | ||||
XM_006723574.3 | Intron | XP_006723637.1 | ||||
XM_006723575.3 | Intron | XP_006723638.1 | ||||
XM_006723577.2 | Intron | XP_006723640.1 | ||||
XM_011529255.2 | Intron | XP_011527557.2 | ||||
XM_011529257.1 | Intron | XP_011527559.1 | ||||
XM_011529258.2 | Intron | XP_011527560.1 | ||||
XM_011529259.2 | Intron | XP_011527561.1 | ||||
XM_017027887.1 | Intron | XP_016883376.1 | ||||
XM_017027888.1 | Intron | XP_016883377.1 | ||||
XM_017027889.1 | Intron | XP_016883378.1 | ||||
XM_017027890.1 | Intron | XP_016883379.1 | ||||
XM_017027891.1 | Intron | XP_016883380.1 | ||||
XM_017027892.1 | Intron | XP_016883381.1 | ||||
XM_017027893.1 | Intron | XP_016883382.1 |
Set Membership: |
HapMap |