Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ESF1 - ESF1 nucleolar pre-rRNA processing protein homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFAF5 - NADH:ubiquinone oxidoreductase complex assembly factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039375.2 | Intron | NP_001034464.1 | ||||
NM_024120.4 | Intron | NP_077025.2 | ||||
XM_006723620.3 | Intron | XP_006723683.1 | ||||
XM_006723622.2 | Intron | XP_006723685.1 | ||||
XM_006723623.2 | Intron | XP_006723686.1 | ||||
XM_006723624.2 | Intron | XP_006723687.1 | ||||
XM_011529342.2 | Intron | XP_011527644.1 | ||||
XM_011529344.2 | Intron | XP_011527646.1 | ||||
XM_017028061.1 | Intron | XP_016883550.1 |
Set Membership: |
HapMap |