Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300611 MIM: 300587 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1GALT1C1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1GALT1C1 - C1GALT1 specific chaperone 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCTS1 - MCTS1, re-initiation and release factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137554.1 | 7464 | UTR 3 | NP_001131026.1 | |||
NM_014060.2 | 7464 | UTR 3 | NP_054779.1 |
Set Membership: |
HapMap |