Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300304 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CUL4B - cullin 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079872.1 | Intron | NP_001073341.1 | ||||
NM_003588.3 | Intron | NP_003579.3 | ||||
XM_005262481.1 | Intron | XP_005262538.1 | ||||
XM_006724784.1 | Intron | XP_006724847.1 | ||||
XM_006724785.2 | Intron | XP_006724848.1 | ||||
XM_011531399.2 | Intron | XP_011529701.1 | ||||
XM_011531400.2 | Intron | XP_011529702.1 | ||||
XM_011531401.1 | Intron | XP_011529703.1 |
Set Membership: |
HapMap |