Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGTGCATATGCACTGGCTTTCC[C/T]TCCACAGTTCCTCTGCAGCTTCGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608618 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM3C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM3C - family with sequence similarity 3 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040020.1 | Intron | NP_001035109.1 | ||||
NM_014888.2 | Intron | NP_055703.1 | ||||
XM_011515736.2 | Intron | XP_011514038.1 | ||||
XM_011515737.2 | Intron | XP_011514039.1 |
Set Membership: |
HapMap |