Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCTTTCAGCCCAGACCTTTGTG[C/G]GGCAAGTGTTGGTAAGTACATTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615215 | ||||||||||||||||||||
Literature Links: |
KCNU1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNU1 - potassium calcium-activated channel subfamily U member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031836.2 | 391 | Missense Mutation | CGG,GGG | R,G 102 | NP_001027006.2 | |
XM_011544424.1 | 391 | Missense Mutation | CGG,GGG | R,G 102 | XP_011542726.1 | |
XM_011544426.1 | 391 | Missense Mutation | CGG,GGG | R,G 102 | XP_011542728.1 | |
XM_011544427.1 | 391 | Missense Mutation | CGG,GGG | R,G 102 | XP_011542729.1 | |
XM_017013144.1 | 391 | Missense Mutation | CGG,GGG | R,G 102 | XP_016868633.1 |