Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTGCATGCAGACTGCCTGCTTG[A/G]GAAACATACTTCTTTATATGCCCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609326 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf166-AS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C20orf166-AS1 - C20orf166 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1-1 - microRNA 1-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1-1HG - MIR1-1 host gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302812.1 | Intron | NP_001289741.1 | ||||
NM_178463.3 | Intron | NP_848558.1 |