Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCATTTTTATAAACTTTCCTAAA[A/G]GCTCAGAGAACAAGGTGTATTTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612471 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGXT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AGXT2 - alanine--glyoxylate aminotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306173.1 | Intron | NP_001293102.1 | ||||
NM_031900.3 | Intron | NP_114106.1 | ||||
XM_005248337.2 | Intron | XP_005248394.1 | ||||
XM_005248338.2 | Intron | XP_005248395.1 | ||||
XM_017009748.1 | Intron | XP_016865237.1 |