Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAGCAGTTCAATTAATTTTCAC[C/T]GGAAAATAATATTCTAATTGTGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612396 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALLC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALLC - allantoicase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DCDC2C - doublecortin domain containing 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287444.1 | Intron | NP_001274373.1 | ||||
XM_006711894.3 | Intron | XP_006711957.1 | ||||
XM_011510393.2 | Intron | XP_011508695.1 | ||||
XM_011510394.2 | Intron | XP_011508696.1 | ||||
XM_017004836.1 | Intron | XP_016860325.1 | ||||
XM_017004837.1 | Intron | XP_016860326.1 | ||||
XM_017004838.1 | Intron | XP_016860327.1 |