Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604822 | ||||||||||||||||||||
Literature Links: |
CAPN11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN11 - calpain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007058.3 | Intron | NP_008989.2 | ||||
XM_006714984.1 | Intron | XP_006715047.1 | ||||
XM_006714985.1 | Intron | XP_006715048.1 | ||||
XM_006714987.1 | Intron | XP_006715050.1 | ||||
XM_011514274.1 | Intron | XP_011512576.1 | ||||
XM_011514275.1 | Intron | XP_011512577.1 |