Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTTTGAGGACGCTCGTGATTA[C/T]ATTGGCCCACCTAGATAATCCAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602108 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MATN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MATN2 - matrilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317748.1 | Intron | NP_001304677.1 | ||||
NM_002380.4 | Intron | NP_002371.3 | ||||
NM_030583.3 | Intron | NP_085072.2 | ||||
XM_005250920.1 | Intron | XP_005250977.1 | ||||
XM_017013417.1 | Intron | XP_016868906.1 | ||||
XM_017013418.1 | Intron | XP_016868907.1 |
Set Membership: |
HapMap |