Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEND6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEND6 - BEN domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318539.1 | Intron | NP_001305468.1 | ||||
NM_152731.2 | Intron | NP_689944.2 | ||||
XM_005248889.1 | Intron | XP_005248946.1 | ||||
XM_011514346.2 | Intron | XP_011512648.1 | ||||
XM_017010406.1 | Intron | XP_016865895.1 | ||||
XM_017010407.1 | Intron | XP_016865896.1 | ||||
XM_017010408.1 | Intron | XP_016865897.1 |
DST - dystonin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |