Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F3 - E2F transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243076.2 | Intron | NP_001230005.1 | ||||
NM_001949.4 | Intron | NP_001940.1 | ||||
XM_005248865.4 | Intron | XP_005248922.1 | ||||
XM_005248866.4 | Intron | XP_005248923.1 | ||||
XM_011514324.2 | Intron | XP_011512626.1 | ||||
XM_011514328.2 | Intron | XP_011512630.1 | ||||
XM_017010330.1 | Intron | XP_016865819.1 |