Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAGATGGTCACTTCTGAGAAGG[A/G]CAAAACAAAAAATAGATGCCATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615787 MIM: 601436 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NADK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NADK2 - NAD kinase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085411.2 | Intron | NP_001078880.1 | ||||
NM_001287340.1 | Intron | NP_001274269.1 | ||||
NM_001287341.1 | Intron | NP_001274270.1 | ||||
NM_153013.4 | Intron | NP_694558.1 | ||||
XM_005248241.4 | Intron | XP_005248298.1 |
SKP2 - S-phase kinase-associated protein 2, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |