Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151740 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANXA2 - annexin A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002857.1 | Intron | NP_001002857.1 | ||||
NM_001002858.2 | Intron | NP_001002858.1 | ||||
NM_001136015.2 | Intron | NP_001129487.1 | ||||
NM_004039.2 | Intron | NP_004030.1 | ||||
XM_011521477.2 | Intron | XP_011519779.1 | ||||
XM_017022090.1 | Intron | XP_016877579.1 | ||||
XM_017022091.1 | Intron | XP_016877580.1 | ||||
XM_017022092.1 | Intron | XP_016877581.1 | ||||
XM_017022093.1 | Intron | XP_016877582.1 |
Set Membership: |
HapMap |