Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATGAGCCTGGGCCTGACTGCA[A/G]GCTCCTGTTGGGAGCTCGCGGAGGT
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 608833 MIM: 608362 | ||||||||||||||||||||||||||
Literature Links: |
RTEL1 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
RTEL1 - regulator of telomere elongation helicase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTEL1-TNFRSF6B - RTEL1-TNFRSF6B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STMN3 - stathmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276310.1 | Intron | NP_001263239.1 | ||||
NM_015894.3 | Intron | NP_056978.2 |