Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612243 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGRG6 - adhesion G protein-coupled receptor G6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032394.2 | Intron | NP_001027566.1 | ||||
NM_001032395.2 | Intron | NP_001027567.1 | ||||
NM_020455.5 | Intron | NP_065188.4 | ||||
NM_198569.2 | Intron | NP_940971.1 | ||||
XM_005267061.3 | Intron | XP_005267118.1 | ||||
XM_006715516.2 | Intron | XP_006715579.1 | ||||
XM_006715517.2 | Intron | XP_006715580.1 | ||||
XM_006715518.2 | Intron | XP_006715581.1 | ||||
XM_011535964.1 | Intron | XP_011534266.1 | ||||
XM_017011085.1 | Intron | XP_016866574.1 |
Set Membership: |
HapMap |