Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTATGCTGGTAAACCCTTAACTC[A/G]GTATGTATCAAGAATGCTGGCCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611617 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFHD1 - EF-hand domain family member D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243252.1 | Intron | NP_001230181.1 | ||||
NM_001308395.1 | Intron | NP_001295324.1 | ||||
NM_025202.3 | Intron | NP_079478.1 | ||||
XM_011511924.2 | Intron | XP_011510226.1 |
Set Membership: |
HapMap |