Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600489 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFATC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NFATC1 - nuclear factor of activated T-cells 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278669.1 | Intron | NP_001265598.1 | ||||
NM_001278670.1 | Intron | NP_001265599.1 | ||||
NM_001278672.1 | Intron | NP_001265601.1 | ||||
NM_001278673.1 | Intron | NP_001265602.1 | ||||
NM_001278675.1 | Intron | NP_001265604.1 | ||||
NM_006162.4 | Intron | NP_006153.2 | ||||
NM_172387.2 | Intron | NP_765975.1 | ||||
NM_172388.2 | Intron | NP_765976.1 | ||||
NM_172389.2 | Intron | NP_765977.1 | ||||
NM_172390.2 | Intron | NP_765978.1 | ||||
XM_017025783.1 | Intron | XP_016881272.1 |
Set Membership: |
HapMap |