Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCCACACACAACTATGTTGTTC[A/G]ACTATTAGCCACAAATTTGCTCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164343 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EBF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EBF1 - early B-cell factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290360.2 | Intron | NP_001277289.1 | ||||
NM_001324101.1 | Intron | NP_001311030.1 | ||||
NM_001324103.1 | Intron | NP_001311032.1 | ||||
NM_001324106.1 | Intron | NP_001311035.1 | ||||
NM_001324107.1 | Intron | NP_001311036.1 | ||||
NM_001324108.1 | Intron | NP_001311037.1 | ||||
NM_001324109.1 | Intron | NP_001311038.1 | ||||
NM_001324111.1 | Intron | NP_001311040.1 | ||||
NM_024007.4 | Intron | NP_076870.1 | ||||
NM_182708.2 | Intron | NP_874367.1 | ||||
XM_017009192.1 | Intron | XP_016864681.1 | ||||
XM_017009193.1 | Intron | XP_016864682.1 | ||||
XM_017009194.1 | Intron | XP_016864683.1 | ||||
XM_017009195.1 | Intron | XP_016864684.1 | ||||
XM_017009196.1 | Intron | XP_016864685.1 | ||||
XM_017009197.1 | Intron | XP_016864686.1 | ||||
XM_017009198.1 | Intron | XP_016864687.1 | ||||
XM_017009199.1 | Intron | XP_016864688.1 | ||||
XM_017009200.1 | Intron | XP_016864689.1 | ||||
XM_017009201.1 | Intron | XP_016864690.1 | ||||
XM_017009202.1 | Intron | XP_016864691.1 | ||||
XM_017009203.1 | Intron | XP_016864692.1 | ||||
XM_017009204.1 | Intron | XP_016864693.1 |
Set Membership: |
HapMap |