Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600490 | ||||||||||||||||||||
Literature Links: |
NFATC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NFATC2 - nuclear factor of activated T-cells 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136021.2 | 5977 | UTR 3 | NP_001129493.1 | |||
NM_001258292.1 | 5977 | UTR 3 | NP_001245221.1 | |||
NM_001258294.1 | 5977 | UTR 3 | NP_001245223.1 | |||
NM_001258295.1 | 5977 | UTR 3 | NP_001245224.1 | |||
NM_001258296.1 | 5977 | UTR 3 | NP_001245225.1 | |||
NM_001258297.1 | 5977 | UTR 3 | NP_001245226.1 | |||
NM_012340.4 | 5977 | UTR 3 | NP_036472.2 | |||
NM_173091.3 | 5977 | UTR 3 | NP_775114.1 | |||
XM_011528824.2 | 5977 | Intron | XP_011527126.1 | |||
XM_011528825.2 | 5977 | Intron | XP_011527127.1 | |||
XM_011528826.2 | 5977 | Intron | XP_011527128.1 | |||
XM_017027850.1 | 5977 | Intron | XP_016883339.1 | |||
XM_017027851.1 | 5977 | UTR 3 | XP_016883340.1 |