Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609434 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LUC7L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LUC7L3 - LUC7 like 3 pre-mRNA splicing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006107.3 | Intron | NP_006098.2 | ||||
NM_016424.4 | Intron | NP_057508.2 | ||||
XM_005257448.1 | Intron | XP_005257505.1 | ||||
XM_005257449.2 | Intron | XP_005257506.1 | ||||
XM_005257452.1 | Intron | XP_005257509.1 | ||||
XM_006721943.2 | Intron | XP_006722006.1 |