Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612765 | ||||||||||||||||||||
Literature Links: |
SFI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SFI1 - SFI1 centrin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007467.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | NP_001007468.1 | |
NM_001258325.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | NP_001245254.1 | |
NM_001258326.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | NP_001245255.1 | |
NM_001258327.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | NP_001245256.1 | |
NM_014775.3 | 565 | Missense Mutation | CAC,CTC | H,L 13 | NP_055590.2 | |
XM_005261868.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_005261925.1 | |
XM_006724390.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_006724453.1 | |
XM_011530570.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528872.1 | |
XM_011530571.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528873.1 | |
XM_011530574.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528876.1 | |
XM_011530575.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528877.1 | |
XM_011530576.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528878.1 | |
XM_011530577.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528879.1 | |
XM_011530579.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528881.1 | |
XM_011530580.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528882.1 | |
XM_011530581.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528883.1 | |
XM_011530582.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528884.1 | |
XM_011530583.2 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_011528885.1 | |
XM_011530587.2 | 565 | Intron | XP_011528889.1 | |||
XM_017029139.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884628.1 | |
XM_017029140.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884629.1 | |
XM_017029141.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884630.1 | |
XM_017029142.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884631.1 | |
XM_017029143.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884632.1 | |
XM_017029144.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884633.1 | |
XM_017029145.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884634.1 | |
XM_017029146.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884635.1 | |
XM_017029147.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884636.1 | |
XM_017029148.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884637.1 | |
XM_017029149.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884638.1 | |
XM_017029150.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884639.1 | |
XM_017029151.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884640.1 | |
XM_017029152.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884641.1 | |
XM_017029153.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884642.1 | |
XM_017029154.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884643.1 | |
XM_017029155.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884644.1 | |
XM_017029156.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884645.1 | |
XM_017029157.1 | 565 | Intron | XP_016884646.1 | |||
XM_017029158.1 | 565 | Missense Mutation | CAC,CTC | H,L 13 | XP_016884647.1 | |
XM_017029159.1 | 565 | UTR 5 | XP_016884648.1 | |||
XM_017029160.1 | 565 | Intron | XP_016884649.1 | |||
XM_017029161.1 | 565 | UTR 5 | XP_016884650.1 | |||
XM_017029162.1 | 565 | UTR 5 | XP_016884651.1 |