Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGCTGTGATGGACAATTCACCC[C/T]TTGGTTAGTTACGGACATAATTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606941 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALG9 - ALG9, alpha-1,2-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077690.1 | Intron | NP_001071158.1 | ||||
NM_001077691.1 | Intron | NP_001071159.1 | ||||
NM_001077692.1 | Intron | NP_001071160.1 | ||||
NM_024740.2 | Intron | NP_079016.2 | ||||
XM_005277723.4 | Intron | XP_005277780.1 | ||||
XM_005277724.4 | Intron | XP_005277781.1 | ||||
XM_006718913.2 | Intron | XP_006718976.1 | ||||
XM_011542992.1 | Intron | XP_011541294.1 | ||||
XM_017018313.1 | Intron | XP_016873802.1 | ||||
XM_017018314.1 | Intron | XP_016873803.1 | ||||
XM_017018315.1 | Intron | XP_016873804.1 | ||||
XM_017018316.1 | Intron | XP_016873805.1 | ||||
XM_017018317.1 | Intron | XP_016873806.1 | ||||
XM_017018318.1 | Intron | XP_016873807.1 | ||||
XM_017018319.1 | Intron | XP_016873808.1 | ||||
XM_017018320.1 | Intron | XP_016873809.1 | ||||
XM_017018321.1 | Intron | XP_016873810.1 | ||||
XM_017018322.1 | Intron | XP_016873811.1 | ||||
XM_017018323.1 | Intron | XP_016873812.1 | ||||
XM_017018324.1 | Intron | XP_016873813.1 | ||||
XM_017018325.1 | Intron | XP_016873814.1 | ||||
XM_017018326.1 | Intron | XP_016873815.1 |
Set Membership: |
HapMap |