Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGN3 - high mobility group nucleosomal binding domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201362.1 | Intron | NP_001188291.1 | ||||
NM_001201363.1 | Intron | NP_001188292.1 | ||||
NM_001318884.1 | Intron | NP_001305813.1 | ||||
NM_001318885.1 | Intron | NP_001305814.1 | ||||
NM_001318886.1 | Intron | NP_001305815.1 | ||||
NM_001318887.1 | Intron | NP_001305816.1 | ||||
NM_001318888.1 | Intron | NP_001305817.1 | ||||
NM_004242.3 | Intron | NP_004233.1 | ||||
NM_138730.2 | Intron | NP_620058.1 |