Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCACTTTTGGGCTTCACAGCGCC[A/G]AGTTCCTATAAAACTGTTGAGTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600267 | ||||||||||||||||||||
Literature Links: |
PTPN13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPN13 - protein tyrosine phosphatase, non-receptor type 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006264.2 | Intron | NP_006255.1 | ||||
NM_080683.2 | Intron | NP_542414.1 | ||||
NM_080684.2 | Intron | NP_542415.1 | ||||
NM_080685.2 | Intron | NP_542416.1 | ||||
XM_011532165.1 | Intron | XP_011530467.1 | ||||
XM_017008511.1 | Intron | XP_016864000.1 | ||||
XM_017008512.1 | Intron | XP_016864001.1 | ||||
XM_017008513.1 | Intron | XP_016864002.1 | ||||
XM_017008514.1 | Intron | XP_016864003.1 | ||||
XM_017008515.1 | Intron | XP_016864004.1 | ||||
XM_017008516.1 | Intron | XP_016864005.1 |