Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACAGTCACAGGTTTTAAAGGC[A/T]TTTCTCTTTTTCTGCGGCTTCTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612316 MIM: 612317 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD3A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATAD3A - ATPase family, AAA domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170535.1 | Intron | NP_001164006.1 | ||||
NM_001170536.1 | Intron | NP_001164007.1 | ||||
NM_018188.3 | Intron | NP_060658.3 | ||||
XM_017001628.1 | Intron | XP_016857117.1 |
ATAD3B - ATPase family, AAA domain containing 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |