Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACACTGATCATCTCTTTGTGTTG[G/T]CAATATTCAAAATTCTCACTTTTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR70 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
WDR70 - WD repeat domain 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018034.2 | Intron | NP_060504.1 | ||||
XM_005248318.3 | Intron | XP_005248375.1 | ||||
XM_011514061.2 | Intron | XP_011512363.1 | ||||
XM_017009604.1 | Intron | XP_016865093.1 | ||||
XM_017009605.1 | Intron | XP_016865094.1 |