Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTATGTCTCGCTAAAGAGTCTG[C/G]ACTTCGTATTGTGGGCAGTAGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607005 MIM: 611835 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GEMIN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GEMIN5 - gem nuclear organelle associated protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL22 - mitochondrial ribosomal protein L22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014990.2 | Intron | NP_001014990.1 | ||||
NM_014180.3 | Intron | NP_054899.2 |
Set Membership: |
HapMap |