Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTCACTCTAAGGAGTTTTAACT[A/C]TGTAAGAAATTAAAAACCAAAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604281 MIM: 610821 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DBF4 - DBF4 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318060.1 | Intron | NP_001304989.1 | ||||
NM_001318061.1 | Intron | NP_001304990.1 | ||||
NM_001318062.1 | Intron | NP_001304991.1 | ||||
NM_006716.3 | Intron | NP_006707.1 |
SLC25A40 - solute carrier family 25 member 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |