Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACATTATGAAAACCTGGAATTT[C/T]ACAATGTAATCTTAGAATCTGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609232 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102467212 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102467212 - uncharacterized LOC102467212 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT12 - nudix hydrolase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300741.1 | 3164 | UTR 3 | NP_001287670.1 | |||
NM_031438.3 | 3164 | UTR 3 | NP_113626.1 | |||
XM_005272095.1 | 3164 | UTR 3 | XP_005272152.1 | |||
XM_005272097.3 | 3164 | UTR 3 | XP_005272154.1 |
Set Membership: |
HapMap |